6-Brom-2-(cyclopropylamino)pyridin
Lieferant: COMBI-BLOCKS
HC-2588-1G
HC-2588-5G
COBBHC-2588-1GEA
149
EUR
COBBHC-2588-1G
COBBHC-2588-5G
6-Brom-2-(cyclopropylamino)pyridin
6-Brom-2-(cyclopropylamino)pyridin
Formel:
C₈H₉BrN₂ |
MDL:
MFCD09878950 CAS-Nummer: 959237-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...