5-Brom-2-(cyclopropylamino)-3-nitropyridin

Lieferant: COMBI-BLOCKS

PY-7330-1G PY-7330-5G
COBBPY-7330-1GEA 98.3 EUR
COBBPY-7330-1G COBBPY-7330-5G
5-Brom-2-(cyclopropylamino)-3-nitropyridin
5-Brom-2-(cyclopropylamino)-3-nitropyridin
Formel: C₈H₈BrN₃O₂
MDL: MFCD08692115
CAS-Nummer: 1010422-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR