N-tert-Butyl-2-chlorisonicotinamid
Lieferant: COMBI-BLOCKS
PY-1297-5G
PY-1297-1G
COBBPY-1297-5GEA
451
EUR
COBBPY-1297-5G
COBBPY-1297-1G
N-tert-Butyl-2-chlorisonicotinamid
N-tert-Butyl-2-chlorisonicotinamid
Formel:
C₁₀H₁₃ClN₂O |
MDL:
MFCD11128609 CAS-Nummer: 588694-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...