2-Chlor-N-methylisonicotinamid

Lieferant: COMBI-BLOCKS

Synonyme: N-Methyl 2-chloroisonicotinamide

PY-7942-5G PY-7942-1G
COBBPY-7942-5GEA 759 EUR
COBBPY-7942-5G COBBPY-7942-1G
2-Chlor-N-methylisonicotinamid
2-Chlor-N-methylisonicotinamid
Formel: C₇H₇ClN₂O
Molecular Weight: 170.6000 g/mol
MDL: MFCD09862946
CAS-Nummer: 131418-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR