2-(Cyclohexylamino)nicotinonitril

Lieferant: COMBI-BLOCKS

PY-7986-1G PY-7986-5G
COBBPY-7986-1GEA 149 EUR
COBBPY-7986-1G COBBPY-7986-5G
2-(Cyclohexylamino)nicotinonitril
2-(Cyclohexylamino)nicotinonitril
Formel: C₁₂H₁₅N₃
MDL: MFCD09426708
CAS-Nummer: 77276-34-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR