2-(Cyclopropylamino)nicotinonitril

Lieferant: COMBI-BLOCKS

PY-7987-1G PY-7987-5G
COBBPY-7987-1GEA 149 EUR
COBBPY-7987-1G COBBPY-7987-5G
2-(Cyclopropylamino)nicotinonitril
2-(Cyclopropylamino)nicotinonitril
Formel: C₉H₉N₃
Molecular Weight: 159,190 g/mol
MDL: MFCD09455268
CAS-Nummer: 52583-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR