2-Ethoxynicotinonitril

Lieferant: COMBI-BLOCKS

CA-4238-5G CA-4238-1G
COBBCA-4238-5GEA 497 EUR
COBBCA-4238-5G COBBCA-4238-1G
2-Ethoxynicotinonitril
2-Ethoxynicotinonitril
Formel: C₇H₆N₂O
MDL: MFCD08436064
CAS-Nummer: 14248-71-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR