6-Methoxynicotinonitril

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxypyridine-5-carbonitrile

PY-7018-1G PY-7018-25G PY-7018-5G
COBBPY-7018-1GEA 30.8 EUR
COBBPY-7018-1G COBBPY-7018-25G COBBPY-7018-5G
6-Methoxynicotinonitril
6-Methoxynicotinonitril
Formel: C₇H₆N₂O
Molecular Weight: 134.1400 g/mol
MDL: MFCD00084981
CAS-Nummer: 15871-85-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR