3-Methyl-4-pyridinboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

BB-8503-1G BB-8503-250M
COBBBB-8503-1GEA 759 EUR
COBBBB-8503-1G COBBBB-8503-250M
3-Methyl-4-pyridinboronsäure Hydrochlorid
3-Methyl-4-pyridinboronsäure Hydrochlorid
Formel: C₆H₉BClNO₂
MDL: MFCD06659519
CAS-Nummer: 1072952-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR