6-Chlor-4-(trifluormethyl)-2-pyridinboronsäure
Lieferant: COMBI-BLOCKS
BB-5875-1G
BB-5875-250M
COBBBB-5875-1GEA
0
EUR
COBBBB-5875-1G
COBBBB-5875-250M
6-Chlor-4-(trifluormethyl)-2-pyridinboronsäure
6-Chloro-4-(trifluoromethyl)-2-pyridineboronic acid
Formel:
C₆H₄BClF₃NO₂ |
MDL:
MFCD12546508 CAS-Nummer: 1217500-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...