6-Chlor-4-(trifluormethyl)-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
BB-5860-1G
BB-5860-250M
COBBBB-5860-1GEA
1430
EUR
COBBBB-5860-1G
COBBBB-5860-250M
6-Chlor-4-(trifluormethyl)-3-pyridinboronsäure
6-Chloro-4-(trifluoromethyl)-3-pyridineboronic acid
Formel:
C₆H₄BClF₃NO₂ |
MDL:
MFCD12546506 CAS-Nummer: 1217500-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...