2-Amino-4-(trifluormethyl)-5-pyridinboronsäurepinakolester 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridin-2-amine, 2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester
H54372.MD
H54372.03
H54372.MDEA
192
EUR
H54372.MD
H54372.03
2-Amino-4-(trifluormethyl)-5-pyridinboronsäurepinakolester 95%
2-Amino-4-(trifluormethyl)-5-pyridinboronsäurepinakolester
Purity: 95%
CAS No.: 944401-57-4
Molecular Formula: C12H16BF3N2O2
Formula Weight: 288.07
Beilstein Reference: 15519102
MDL No.: MFCD12923420
Formel:
C₁₂H₁₆BF₃N₂O₂ Molecular Weight: 288.07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12923420 CAS-Nummer: 944401-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...