2-(Benzylamino)-5-pyridinboronsäurepinakolester 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-(Benzylamino)pyridine-3-boronic acid pinacol ester, N-Benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
H27463.MD
H27463.03
H27463.MDEA
73.3
EUR
H27463.MD
H27463.03
2-(Benzylamino)-5-pyridinboronsäurepinakolester 95%
2-(Benzylamino)-5-pyridinboronsäurepinakolester
Purity: 95%
CAS No.: 1073354-27-4
Molecular Formula: C18H23BN2O2
Formula Weight: 310.20
Melting Point: 135-138°
MDL No.: MFCD06798270
Formel:
C₁₈H₂₃BN₂O₂ Molecular Weight: 310,20 g/mol Schmelzpunkt: 135…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06798270 CAS-Nummer: 1073354-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...