2-(4-Formylphenyl)isonicotinonitril

Lieferant: BIONET RESEARCH

3Y-0716.1 3Y-0716.10 3Y-0716.5 3Y-0716.500
BIOT3Y-0716.1EA 0 EUR
BIOT3Y-0716.1 BIOT3Y-0716.10 BIOT3Y-0716.5 BIOT3Y-0716.500
2-(4-Formylphenyl)isonicotinonitril
2-(4-Formylphenyl)isonicotinonitril
Formel: C₁₃H₈N₂O
MDL: MFCD06200959
CAS-Nummer: 253801-11-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR