6-(4-Fluorphenyl)-3-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(4-Fluorophenyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-(4-Fluorophenyl)pyridine-3-boronic acid pinacol ester
PN-8778-1G
PN-8778-250M
COBBPN-8778-1GEA
748
EUR
COBBPN-8778-1G
COBBPN-8778-250M
6-(4-Fluorphenyl)-3-pyridinboronsäurepinakolester
6-(4-Fluorphenyl)-3-pyridinboronsäurepinakolester
Formel:
C₁₇H₁₉BFNO₂ Molecular Weight: 299.15 g/mol |
MDL:
MFCD09037494 CAS-Nummer: 1073354-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...