5-(1-Chlor-2-methylpropan-2-yl)-3-(4-methylphenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6527-5G HC-6527-1G
COBBHC-6527-5GEA 595 EUR
COBBHC-6527-5G COBBHC-6527-1G
5-(1-Chlor-2-methylpropan-2-yl)-3-(4-methylphenyl)-1,2,4-oxadiazol
5-(1-Chlor-2-methylpropan-2-yl)-3-(4-methylphenyl)-1,2,4-oxadiazol
Formel: C₁₃H₁₅ClN₂O
MDL: MFCD11504918
CAS-Nummer: 1072944-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR