5-(Dichlormethyl)-3-(4-methylphenyl)-1,2,4-oxadiazol
Lieferant: COMBI-BLOCKS
HC-6555-1G
HC-6555-5G
COBBHC-6555-1GEA
0
EUR
COBBHC-6555-1G
COBBHC-6555-5G
5-(Dichlormethyl)-3-(4-methylphenyl)-1,2,4-oxadiazol
5-(Dichloromethyl)-3-(4-methylphenyl)-1,2,4-oxadiazole
Formel:
C₁₀H₈Cl₂N₂O |
MDL:
MFCD11855936 CAS-Nummer: 59455-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...