5-(1-Chlor-2-methylpropan-2-yl)-3-(3-nitrophenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6536-5G HC-6536-1G
COBBHC-6536-5GEA 595 EUR
COBBHC-6536-5G COBBHC-6536-1G
5-(1-Chlor-2-methylpropan-2-yl)-3-(3-nitrophenyl)-1,2,4-oxadiazol
5-(1-Chlor-2-methylpropan-2-yl)-3-(3-nitrophenyl)-1,2,4-oxadiazol
Formel: C₁₂H₁₂ClN₃O₃
MDL: MFCD11855935
CAS-Nummer: 1133116-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR