5-(Chlormethyl)-3-(3-nitrophenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6532-1G HC-6532-5G
COBBHC-6532-1GEA 98.3 EUR
COBBHC-6532-1G COBBHC-6532-5G
5-(Chlormethyl)-3-(3-nitrophenyl)-1,2,4-oxadiazol
5-(Chloromethyl)-3-(3-nitrophenyl)-1,2,4-oxadiazole
Formel: C₉H₆ClN₃O₃
MDL: MFCD01764743
CAS-Nummer: 6595-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR