5-(Chlormethyl)-3-(4-nitrophenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6522-5G HC-6522-1G
COBBHC-6522-5GEA 876 EUR
COBBHC-6522-5G COBBHC-6522-1G
5-(Chlormethyl)-3-(4-nitrophenyl)-1,2,4-oxadiazol
5-(Chloromethyl)-3-(4-nitrophenyl)-1,2,4-oxadiazole
Formel: C₉H₆ClN₃O₃
MDL: MFCD00452243
CAS-Nummer: 57611-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR