5-(Dichlormethyl)-3-(4-nitrophenyl)-1,2,4-oxadiazol
Lieferant: COMBI-BLOCKS
HC-6523-1G
HC-6523-5G
COBBHC-6523-1GEA
0
EUR
COBBHC-6523-1G
COBBHC-6523-5G
5-(Dichlormethyl)-3-(4-nitrophenyl)-1,2,4-oxadiazol
5-(Dichloromethyl)-3-(4-nitrophenyl)-1,2,4-oxadiazole
Formel:
C₉H₅Cl₂N₃O₃ |
MDL:
MFCD01883262 CAS-Nummer: 905107-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...