5-(Chlormethyl)-3-(4-methoxyphenyl)-1,2,4-oxadiazol

Lieferant: Thermo Scientific

RF01831.250 RF01831.1
MAYBRF01831.250EA 0 EUR
MAYBRF01831.250 MAYBRF01831.1
5-(Chlormethyl)-3-(4-methoxyphenyl)-1,2,4-oxadiazol
5-(Chloromethyl)-3-(4-methoxyphenyl)-1,2,4-oxadiazole
Formel: C₁₀H₉ClN₂O₂
CAS-Nummer: 57238-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR