5-Ethyl-3-(4-methylphenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

Synonyme: 5-Ethyl-3-p-tolyl-1,2,4-oxadiazole

HC-6511-5G HC-6511-1G
COBBHC-6511-5GEA 451 EUR
COBBHC-6511-5G COBBHC-6511-1G
5-Ethyl-3-(4-methylphenyl)-1,2,4-oxadiazol
5-Ethyl-3-(4-methylphenyl)-1,2,4-oxadiazole
Formel: C₁₁H₁₂N₂O
MDL: MFCD09972167
CAS-Nummer: 81386-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR