3-(2-Bromphenyl)-5-(2,3-dichlorphenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6286-1G HC-6286-5G
COBBHC-6286-1GEA 149 EUR
COBBHC-6286-1G COBBHC-6286-5G
3-(2-Bromphenyl)-5-(2,3-dichlorphenyl)-1,2,4-oxadiazol
3-(2-Bromophenyl)-5-(2,3-dichlorophenyl)-1,2,4-oxadiazole
Formel: C₁₄H₇BrCl₂N₂O
MDL: MFCD11053868
CAS-Nummer: 1065074-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR