Ethyl-3-(4-bromphenyl)-1,2,4-oxadiazol-5-carboxylat

Lieferant: COMBI-BLOCKS

HC-6564-5G HC-6564-1G
COBBHC-6564-5GEA 911 EUR
COBBHC-6564-5G COBBHC-6564-1G
Ethyl-3-(4-bromphenyl)-1,2,4-oxadiazol-5-carboxylat
Ethyl-3-(4-bromphenyl)-1,2,4-oxadiazol-5-carboxylat
Formel: C₁₁H₉BrN₂O₃
MDL: MFCD09831978
CAS-Nummer: 861146-12-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR