3-(2-Bromphenyl)-5-(2-fluorphenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6280-5G HC-6280-1G
COBBHC-6280-5GEA 451 EUR
COBBHC-6280-5G COBBHC-6280-1G
3-(2-Bromphenyl)-5-(2-fluorphenyl)-1,2,4-oxadiazol
3-(2-Bromophenyl)-5-(2-fluorophenyl)-1,2,4-oxadiazole
Formel: C₁₄H₈BrFN₂O
MDL: MFCD09878375
CAS-Nummer: 1000339-25-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR