3-(4-Fluorphenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HI-1671-5G HI-1671-1G
COBBHI-1671-5GEA 451 EUR
COBBHI-1671-5G COBBHI-1671-1G
3-(4-Fluorphenyl)-1,2,4-oxadiazol
3-(4-Fluorophenyl)-1,2,4-oxadiazole
Formel: C₈H₅FN₂O
MDL: MFCD16618955
CAS-Nummer: 887763-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR