4-Brom-N-(furan-2-ylmethyl)-3-(trifluormethyl)benzolsulfonamid
Lieferant: COMBI-BLOCKS
HC-6463-5G
HC-6463-1G
COBBHC-6463-5GEA
1180
EUR
COBBHC-6463-5G
COBBHC-6463-1G
4-Brom-N-(furan-2-ylmethyl)-3-(trifluormethyl)benzolsulfonamid
4-Brom-N-(furan-2-ylmethyl)-3-(trifluormethyl)benzolsulfonamid
Formel:
C₁₂H₉BrF₃NO₃S |
MDL:
MFCD09972153 CAS-Nummer: 1020252-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...