3-Brom-N-(furan-2-ylmethyl)-5-methylbenzolsulfonamid
Lieferant: COMBI-BLOCKS
HC-6454-1G
HC-6454-5G
COBBHC-6454-1GEA
149
EUR
COBBHC-6454-1G
COBBHC-6454-5G
3-Brom-N-(furan-2-ylmethyl)-5-methylbenzolsulfonamid
3-Brom-N-(furan-2-ylmethyl)-5-methylbenzolsulfonamid
Formel:
C₁₂H₁₂BrNO₃S |
MDL:
MFCD09972146 CAS-Nummer: 1020252-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...