[3-(4-Chlorphenyl)-5-isoxazolyl]methanamin
Lieferant: BIONET RESEARCH
6M-343S.500
6M-343S.1
6M-343S.5
6M-343S.10
BIOT6M-343S.500EA
0
EUR
BIOT6M-343S.500
BIOT6M-343S.1
BIOT6M-343S.5
BIOT6M-343S.10
[3-(4-Chlorphenyl)-5-isoxazolyl]methanamin
[3-(4-Chlorphenyl)-5-isoxazolyl]methanamin
Formel:
C₁₀H₉ClN₂O |
MDL:
MFCD01935956 CAS-Nummer: 66046-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...