[3-(2-Chlorphenyl)-5-isoxazolyl]methanol

Lieferant: BIONET RESEARCH

Synonyme: 3-(2-Chlorophenyl)-5-(hydroxymethyl)isoxazole, 3-(2-Chlorophenyl)-5-isoxazolemethanol

11L-341S.1 11L-341S.10 11L-341S.5 11L-341S.500
BIOT11L-341S.1EA 0 EUR
BIOT11L-341S.1 BIOT11L-341S.10 BIOT11L-341S.5 BIOT11L-341S.500
[3-(2-Chlorphenyl)-5-isoxazolyl]methanol
[3-(2-Chlorphenyl)-5-isoxazolyl]methanol
Formel: C₁₀H₈ClNO₂
Molecular Weight: 209,6300 g/mol
MDL: MFCD04124289
CAS-Nummer: 438565-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR