[3-(4-Chlorphenyl)-5-isoxazolyl]methanol

Lieferant: BIONET RESEARCH

Synonyme: (3-(4-Chlorophenyl)isoxazol-5-yl)methanol

10L-365S.500 10L-365S.1 10L-365S.5 10L-365S.10
BIOT10L-365S.500EA 0 EUR
BIOT10L-365S.500 BIOT10L-365S.1 BIOT10L-365S.5 BIOT10L-365S.10
[3-(4-Chlorphenyl)-5-isoxazolyl]methanol
[3-(4-Chlorphenyl)-5-isoxazolyl]methanol
Formel: C₁₀H₈ClNO₂
Molecular Weight: 209.6290 g/mol
MDL: MFCD01935957
CAS-Nummer: 206055-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR