[3-(4-Methoxyphenyl)-5-isoxazolyl]methanol

Lieferant: BIONET RESEARCH

11L-326S.5 11L-326S.10 11L-326S.500 11L-326S.1
BIOT11L-326S.5EA 0 EUR
BIOT11L-326S.5 BIOT11L-326S.10 BIOT11L-326S.500 BIOT11L-326S.1
[3-(4-Methoxyphenyl)-5-isoxazolyl]methanol
[3-(4-Methoxyphenyl)-5-isoxazolyl]methanol
Formel: C₁₁H₁₁NO₃
MDL: MFCD04124287
CAS-Nummer: 206055-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR