[3-Phenyl-5-isoxazolyl]methanol

Lieferant: BIONET RESEARCH

Synonyme: (3-Phenyl-5-isoxazolyl)methanol

12L-305S.500 12L-305S.1 12L-305S.5 12L-305S.10
BIOT12L-305S.500EA 0 EUR
BIOT12L-305S.500 BIOT12L-305S.1 BIOT12L-305S.5 BIOT12L-305S.10
[3-Phenyl-5-isoxazolyl]methanol
[3-Phenyl-5-isoxazolyl]methanol
Formel: C₁₀H₉NO₂
Molecular Weight: 175,1840 g/mol
MDL: MFCD01928812
CAS-Nummer: 90924-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR