3-Amino-2,6-dibrompyridin
Lieferant: COMBI-BLOCKS
Synonyme:
2,6-Dibromopyridin-3-amine
PY-7375-1G
PY-7375-25G
PY-7375-5G
COBBPY-7375-1GEA
25.5
EUR
COBBPY-7375-1G
COBBPY-7375-25G
COBBPY-7375-5G
3-Amino-2,6-dibrompyridin
3-Amino-2,6-dibromopyridine
Formel:
C₅H₄Br₂N₂ Molecular Weight: 251,9100 g/mol |
MDL:
MFCD00128862 CAS-Nummer: 39856-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...