3-Amino-6-brom-2-methylpyridin

Lieferant: FLUOROCHEM

Synonyme: 5-Amino-2-bromo-6-methylpyridine, 6-Bromo-2-methylpyridin-3-amine

040412-1G 040412-25G
FLUO040412-1GEA 31.1 EUR
FLUO040412-1G FLUO040412-25G
3-Amino-6-brom-2-methylpyridin
3-Amino-6-bromo-2-methylpyridine
Formel: C₆H₇BrN₂
Molecular Weight: 187.04 g/mol
MDL: MFCD03095086
CAS-Nummer: 126325-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR