2-Chlorchinoxalin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

136301-1G 136301-5G
SIAL136301-1GEA 30.4 EUR
SIAL136301-1G SIAL136301-5G
2-Chlorchinoxalin, Sigma-Aldrich®
2-Chloroquinoxaline
Formel: C₈H₅ClN₂
MDL: MFCD00043907
CAS-Nummer: 1448-87-9
EINECS: 215-905-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR