6,7-Dichlorchinoxalin

Lieferant: COMBI-BLOCKS

PY-7956-1G PY-7956-5G
COBBPY-7956-1GEA 204 EUR
COBBPY-7956-1G COBBPY-7956-5G
6,7-Dichlorchinoxalin
6,7-Dichloroquinoxaline
Formel: C₈H₄Cl₂N₂
Molecular Weight: 199,0368 g/mol
MDL: MFCD00456885
CAS-Nummer: 19853-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR