2,6-Dichlorisonicotinonitril
Lieferant: FLUOROCHEM
Synonyme:
4-Cyano-2,6-dichloropyridine
013934-5G
013934-1G
FLUO013934-5GEA
137
EUR
FLUO013934-5G
FLUO013934-1G
2,6-Dichlorisonicotinonitril
2,6-Dichlorisonicotinonitril
Formel:
C₆H₂Cl₂N₂ Molecular Weight: 172.9995 g/mol |
MDL:
MFCD00173784 CAS-Nummer: 32710-65-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...