3-Chlor-2,6-difluorbenzamid
Lieferant: FLUOROCHEM
005589-1G
005589-5G
FLUO005589-1GEA
48.8
EUR
FLUO005589-1G
FLUO005589-5G
3-Chlor-2,6-difluorbenzamid
3-Chlor-2,6-difluorbenzamid
Formel:
C₇H₄ClF₂NO Molecular Weight: 191.56 g/mol |
MDL:
MFCD01631386 CAS-Nummer: 261762-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...