3-Chlor-2-fluor-5-(trifluormethyl)benzamid

Lieferant: FLUOROCHEM

005677-1G 005677-5G
FLUO005677-1GEA 72.9 EUR
FLUO005677-1G FLUO005677-5G
3-Chlor-2-fluor-5-(trifluormethyl)benzamid
3-Chlor-2-fluor-5-(trifluormethyl)benzamid
Formel: C₈H₄ClF₄NO
MDL: MFCD01631399
CAS-Nummer: 129931-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR