3-(tert-Butoxy)-6-chlorpyridazin

Lieferant: BIONET RESEARCH

4T-0010.500 4T-0010.1
BIOT4T-0010.500EA 0 EUR
BIOT4T-0010.500 BIOT4T-0010.1
3-(tert-Butoxy)-6-chlorpyridazin
3-(tert-Butoxy)-6-chlorpyridazin
Formel: C₈H₁₁ClN₂O
MDL: MFCD03425769
CAS-Nummer: 17321-24-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR