2-Fluor-6-methoxybenzamid ≥98%

Lieferant: Thermo Scientific
Achtung

H32774.MD H32774.03 H32774.06
H32774.MDEA 65.3 EUR
H32774.MD H32774.03 H32774.06
2-Fluor-6-methoxybenzamid ≥98%
2-Fluor-6-methoxybenzamid

Purity: 98% CAS No.: 529512-81-0 Molecular Formula: C8H8FNO2 Formula Weight: 169.15 Melting Point: 190-194° MDL No.: MFCD00277474

Formel: C₈H₈FNO₂
Molecular Weight: 169,15 g/mol
Schmelzpunkt: 190…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00277474
CAS-Nummer: 529512-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR