2-Fluor-6-methoxybenzamid

Lieferant: Thermo Scientific

KM07281.250 KM07281.1
MAYBKM07281.250EA 0 EUR
MAYBKM07281.250 MAYBKM07281.1
2-Fluor-6-methoxybenzamid
2-Fluor-6-methoxybenzamid
Formel: C₈H₈FNO₂
Molecular Weight: 169,15 g/mol
MDL: MFCD00277474
CAS-Nummer: 529512-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR