2,2-Difluor-2-p-tolylessigsäure

Lieferant: COMBI-BLOCKS

HC-2557-1G HC-2557-5G
COBBHC-2557-1GEA 595 EUR
COBBHC-2557-1G COBBHC-2557-5G
2,2-Difluor-2-p-tolylessigsäure
2,2-Difluor-2-p-tolylessigsäure
Formel: C₉H₈F₂O₂
MDL: MFCD11007728
CAS-Nummer: 131323-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR