3-Amino-2,2-dimethyl-1-propanol 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L10429.03
L10429.06
L10429.03EA
30.7
EUR
L10429.03
L10429.06
3-Amino-2,2-dimethyl-1-propanol 95%
3-Amino-2,2-dimethyl-1-propanol
Purity: 95%
CAS No.: 26734-09-8
Molecular Formula: C5H13NO
Formula Weight: 103.17
Melting Point: ca 70°
Beilstein Reference: 1731900
Formel:
C₅H₁₃NO Molecular Weight: 103.17 g/mol Schmelzpunkt: ∼ 70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059847 CAS-Nummer: 26734-09-8 EINECS: 247-948-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...