2,2':6',2''-Terpyridin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
TERPY
A17750.01
A17750.03
A17750.06
A17750.01EA
64.5
EUR
A17750.01
A17750.03
A17750.06
2,2':6',2''-Terpyridin ≥97%
2,2':6',2''-Terpyridin
Purity: 97%
CAS No.: 1148-79-4
Molecular Formula: C15H11N3
Formula Weight: 233.28
Physical Form: Powder
Odour: Odourless
UN No.: 2811
MDL No.: MFCD00006213
Formel:
C₁₅H₁₁N₃ Schmelzpunkt: 89…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006213 CAS-Nummer: 1148-79-4 EINECS: 214-559-5 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...