Methyl-3-cyanbenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 3-cyanobenzoate
CA-4667-5G
CA-4667-25G
CA-4667-1G
COBBCA-4667-5GEA
36.2
EUR
COBBCA-4667-5G
COBBCA-4667-25G
COBBCA-4667-1G
Methyl-3-cyanbenzoat
Methyl-3-cyanbenzoat
Formel:
C₉H₇NO₂ Molecular Weight: 161.1574 g/mol |
MDL:
MFCD00220156 CAS-Nummer: 13531-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...