3-Chlor-2-methylbenzonitril

Lieferant: Thermo Scientific

TL00462.1 TL00462.10 TL00462.50
MAYBTL00462.1EA 0 EUR
MAYBTL00462.1 MAYBTL00462.10 MAYBTL00462.50
3-Chlor-2-methylbenzonitril
3-Chlor-2-methylbenzonitril
Formel: C₈H₆ClN
MDL: MFCD00045598
CAS-Nummer: 54454-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR