5-Chlor-2-methylbenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

696358-25G 696358-5G
SIAL696358-25GEA 340 EUR
SIAL696358-25G SIAL696358-5G
5-Chlor-2-methylbenzonitril, Sigma-Aldrich®
5-Chlor-2-methylbenzonitril
Formel: C₈H₆ClN
Molecular Weight: 151,5929 g/mol
MDL: MFCD00045599
CAS-Nummer: 50712-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR